missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Polygoprep™ 60 to 50
203.00 €
Specifications
Particle Size | 40 to 63 μm |
---|---|
Pore Size | 60 Å |
Stationary Phase | POLYGOPREP 60 to 50 |
Technique | Liquid Chromatography |
pH | 2 to 8 |
Specifications
40 to 63 μm | |
POLYGOPREP 60 to 50 | |
2 to 8 | |
350 m2/g | |
Silica |
60 Å | |
Liquid Chromatography | |
POLYGOPREP | |
Polygoprep | |
Liquid Chromatography |