missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Polygoprep™ 60 to 30 C8
Brand: Macherey-Nagel™ 711470.100
This item is not returnable.
View return policy
Specifications
25 to 40 μm | |
POLYGOPREP 60 to 30 | |
2 to 8 | |
POLYGOPREP C8 | |
Polygoprep | |
Liquid Chromatography |
60 Å | |
Liquid Chromatography | |
100 g | |
350 m2/g | |
Silica |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction