Learn More
Polygalacturonic acid, M.W. 25,000-50,000, Thermo Scientific Chemicals
81.00 € - 803.00 €
Chemical Identifiers
CAS | 25990-10-7 |
---|---|
Molecular Formula | (C6H8O6)n |
Molecular Weight (g/mol) | 194.14 |
MDL Number | MFCD00131972 |
InChI Key | AEMOLEFTQBMNLQ-BKBMJHBISA-N |
Synonym | galacturonan, alpha-d-galactopyranuronic acid, calcium pectate, sodium pectate, alpha-d-galacturonic acid, calcium polygalacturonate, pectate, d-galacturonan, sulfated polygalacturonic acid, galacturonate |
PubChem CID | 445929 |
ChEBI | CHEBI:33885 |
IUPAC Name | (2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid |
SMILES | O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386836
|
Thermo Scientific Alfa Aesar
L04206.14 |
25 g |
81.00 €
25g |
In Stock
Log in to see stock. |
|||||
11396836
|
Thermo Scientific Alfa Aesar
L04206.22 |
100 g |
237.18 €
100g |
In Stock
Log in to see stock. |
|||||
11306846
|
Thermo Scientific Alfa Aesar
L04206.36 |
500 g |
803.00 €
500g |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
Description
Polygalacturonic acid is reagent which sequesters metal ions and produces gels that can be used for the production of biomolecules. It is used for drug release or to immobilize cells and enzymes . Polygalacturonic acid may be used to identify, differentiate and characterized plant cell wall degrading polygalacturonase(s) from fungi.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25990-10-7 | |
194.14 | |
AEMOLEFTQBMNLQ-BKBMJHBISA-N | |
445929 | |
(2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid |
(C6H8O6)n | |
MFCD00131972 | |
galacturonan, alpha-d-galactopyranuronic acid, calcium pectate, sodium pectate, alpha-d-galacturonic acid, calcium polygalacturonate, pectate, d-galacturonan, sulfated polygalacturonic acid, galacturonate | |
CHEBI:33885 | |
O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O |
Specifications
25990-10-7 | |
MFCD00131972 | |
Soluble in water. | |
O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O | |
194.14 | |
CHEBI:33885 | |
+250° (c=1 in 0.1N NaOH) | |
Polygalacturonic acid |
(C6H8O6)n | |
galacturonan, alpha-d-galactopyranuronic acid, calcium pectate, sodium pectate, alpha-d-galacturonic acid, calcium polygalacturonate, pectate, d-galacturonan, sulfated polygalacturonic acid, galacturonate | |
AEMOLEFTQBMNLQ-BKBMJHBISA-N | |
(2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid | |
445929 | |
25 to 50 kd | |
Hygroscopic |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.