Learn More
Buerkle™ Polyethylene Round All Purpose Boxes
Box for storing, packing, taking samples, sampling and transport of pastes, granules, powders, soil samples and frozen goods (-40 °C up to briefly +95 °C). Ideal use of space, can be stacked. Complete with press-in lid
4.80 € - 5.40 €
Specifications
Color Transparent
Material Polyethylene
Shape Round
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10286363
View Documents
Buerkle™
6210-0250
250 mL
4.80 €
Each
10780773
View Documents
Buerkle™
6210-0500
500 mL
5.40 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • PE, transparent
  • No crevices
  • |PE, transparent
Specifications
Specifications
Transparent
Polyethylene
Round