Learn More
Masterflex™ Polyethylene Elbow Pipe Adapter, pack of 100
Designed for quick and easy assembly without tools
Brand: Masterflex™ 30626-98
This item is not returnable.
View return policy
Description
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Barbed Fitting | |
Not Autoclavable | |
HDPE | |
100 Fittings | |
3/8 in. | |
3/8 in. | |
1 19/32 in. | |
Translucent |
Barb x NPT(M) Connection | |
1/4 in. | |
1 1/2 in. | |
3/8 in. | |
1/4 in. | |
Ethylene Oxide | |
Elbow-shaped |