Learn More
POLA2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21778-1-AP-20UL
 View more versions of this product
Product Code. 16891375
  • 145.00 € / 20µL
Explore more special offers
Description
Description
The protein encoded by this gene is a zinc finger protein that may act as a transcription factor. The encoded protein may be part of a BHC histone deacetylase complex. Translocation of this gene with the fibroblast growth factor receptor-1 gene (FGFR1) results in a fusion gene, which may be a cause of stem cell leukemia lymphoma syndrome (SCLL). Several transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
POLA2
Polyclonal
Unconjugated
POLA2
DNA polymerase alpha subunit B, POLA2
Rabbit
Antigen Affinity Chromatography
RUO
23649
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14181
POLA2
POLA2 Fusion Protein Ag0271
20 μL