missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLA2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21778-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a zinc finger protein that may act as a transcription factor. The encoded protein may be part of a BHC histone deacetylase complex. Translocation of this gene with the fibroblast growth factor receptor-1 gene (FGFR1) results in a fusion gene, which may be a cause of stem cell leukemia lymphoma syndrome (SCLL). Several transcript variants encoding the same protein have been found for this gene.Specifications
POLA2 | |
Polyclonal | |
Unconjugated | |
POLA2 | |
DNA polymerase alpha subunit B, POLA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23649 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14181 | |
POLA2 | |
POLA2 Fusion Protein Ag0271 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction