missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PO43- f(P)-PHOSPHATE solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
54.00 € - 114.00 €
Chemical Identifiers
CAS | 7558-80-7 |
---|---|
Molecular Formula | H2NaO4P |
Molecular Weight (g/mol) | 119.98 |
MDL Number | MFCD00003527,MFCD00146206 |
InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
PubChem CID | 23672064 |
ChEBI | CHEBI:37585 |
SMILES | [Na+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12982175
|
Fisher Chemical
A 037 100 |
100 mL | Bottle |
54.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12992175
|
Fisher Chemical
A 037 250 |
250 mL | Bottle |
77.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12902185
|
Fisher Chemical
A 037 500 |
500 mL | Bottle |
114.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7558-80-7 | |
119.98 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
CHEBI:37585 |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
23672064 | |
[Na+].OP(O)([O-])=O |
Specifications
1g/L | |
Single-component Standard Solution | |
MFCD00003527,MFCD00146206 | |
[Na+].OP(O)([O-])=O | |
119.98 | |
CHEBI:37585 | |
Liquid |
7558-80-7 | |
H2NaO4P | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate | |
23672064 | |
H2O | |
Phosphate |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title