Learn More
SI Analytics™ Connection Cables and Plugs
SI Analytics™ Connection Cables and Plugs are available in various combinations of electrode socket, instrument plug and cable length.
Brand:  SI Analytics™ 285122464
 View more versions of this product
Product Code. 10466165
  • 74.40 € / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Coaxial plug for pH, redox, ammonia and sodium combination electrodes, pH and redox single electrodes as well as reference electrodes in Plus series. The L and LB series plugs are compatible. The LB sockets are matching colors with the BlueLine electrodes
  • Electrode plug for reference electrodes from the predecessor series
  • Plug for conductivity measuring cells with temperature sensor and oxygen cells, for older models
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Coaxial Electrode Plug
L 2 A
2m Coaxial Cable, DIN Instrument Plug
2 m