missing translation for 'onlineSavingsMsg'
Learn More

Platinum wire, 0.25mm (0.01in) dia, For ISA Type R or S Standard Grade Thermocouple, Thermo Scientific Chemicals

101.51 € - 1092.44 €

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11351378
SDS View Documents
Thermo Scientific Alfa Aesar
010288.BU
25 cm
101.51 €
25cm
Please to purchase this item. Need a web account? Register with us today!
11361378
SDS View Documents
Thermo Scientific Alfa Aesar
010288.BY
100 cm
296.48 €
100cm
Please to purchase this item. Need a web account? Register with us today!
11341378
SDS View Documents
Thermo Scientific Alfa Aesar
010288.CC
500 cm
1092.44 €
500cm
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-116-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum wire, 0.25mm (0.01in) dia, For ISA Type R or S Standard Grade Thermocouple, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.