missing translation for 'onlineSavingsMsg'
Learn More

Platinum Wire, 0.076mm (0.003 in.) dia., Thermo Scientific™

Form: Approx .0.045g/50cm

POA

Chemical Identifiers

CAS 7440-6-6
Molecular Formula Pt:Rh; 70:30 wt%
MDL Number MFCD00198186
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346777
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010079.BW
50 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11356777
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010079.G2
2 m This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7440-6-6
Pt:Rh; 70:30 wt%
MFCD00198186

Specifications

Silver
Silver colored wire
MFCD00198186
Wire
7440-6-6
Pt:Rh; 70:30 wt%
Odorless
Platinum, 30 wt% Rhodium wire
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications