Learn More
Platinum wire, 0.0508mm (0.002in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
60.90 € - 704.51 €
Chemical Identifiers
CAS | 7440-06-4 |
---|---|
Molecular Formula | Pt |
Molecular Weight (g/mol) | 195.08 |
MDL Number | MFCD00011179 |
InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
Synonym | black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion |
PubChem CID | 23939 |
ChEBI | CHEBI:33400 |
IUPAC Name | platinum |
SMILES | [Pt] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386848
|
Thermo Scientific Alfa Aesar
013479.G1 |
1 m |
60.90 €
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396848
|
Thermo Scientific Alfa Aesar
013479.G5 |
5 m |
183.92 €
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376848
|
Thermo Scientific Alfa Aesar
013479.G9 |
25 m |
704.51 €
25m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Specifications
7440-06-4 | |
MFCD00011179 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
99.95% | |
≈0.043g/m | |
(metals basis) |
Pt | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
[Pt] | |
195.08 | |
CHEBI:33400 | |
Odorless | |
Wire | |
Platinum wire, 0.0508mm (0.002 in.) dia., Hard, Temper: as drawn |
Safety and Handling
missing translation for 'einecsNumber' : 231-116-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.