Learn More
Dostmann Electronic Platinum Immersion Probe
With a response time of 12 seconds and a temperature range of –200 to 500°, this platinum immersion probe is ideal for many research, scientific, and laboratory environments.
Brand:  Dostmann Electronic 6000-1073
Product Code. 10701654
  • 292.00 € / Each
Explore more special offers
Description
Description
  • Temperature Probe Pt100 6000-1073
  • High-accuracy resistance probe, Pt100 1/10-DIN, Platinium-ceramic sensor IEC 751, 4-wire, cable 1000mm PVC
  • recommend for accurate measurings e.g. with certificate
  • Immersion probe, with handle, mineral-insulated, WS 1.4571, -200..+450°C, 150 x 3.0mm
Specifications
Specifications
12 sec.
-200°C to +500°C
150 mm