missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) bromide, 99.9% (metals basis), Pt 54.5% min, Thermo Scientific Chemicals

Product Code. 11316558
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Platinum(II) bromide used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-12-4
Molecular Formula Br2Pt
Molecular Weight (g/mol) 354.89
MDL Number MFCD00011180
InChI Key KGRJUMGAEQQVFK-UHFFFAOYSA-L
Synonym platinum ii bromide, platinum bromide, platinum bromide ptbr2, platinum dibromide, ptbr2, acmc-1c2hr, platinum bromide, ptbr2
PubChem CID 83486
IUPAC Name dibromoplatinum
SMILES [Br-].[Br-].[Pt++]

Specifications

Melting Point 250°C (decomposition)
Physical Form Crystalline
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 354.91
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 54.5% min.
Assay (metals basis)
Chemical Name or Material Platinum(II) bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.