missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) bromide, 98%, Thermo Scientific Chemicals

Product Code. 10266490
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 250°C
CAS 13455-12-4
Color Brown to Khaki
Physical Form Powder
Quantity 1 g
Assay Percent Range 97.5% min
Molecular Formula Br2Pt
Linear Formula PtBr2
MDL Number MFCD00011180
Synonym platinum ii bromide, platinum bromide, platinum bromide ptbr2, platinum dibromide, ptbr2, acmc-1c2hr, platinum bromide, ptbr2
InChI Key KGRJUMGAEQQVFK-UHFFFAOYSA-L
SMILES [Br-].[Br-].[Pt++]
IUPAC Name dibromoplatinum
Molecular Weight (g/mol) 354.89
PubChem CID 83486
Formula Weight 354.9
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 6.65
Density 6.6500g/mL
Chemical Name or Material Platinum(II) bromide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Platinum(II) bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.