Learn More
Platinum(II) 2,4-pentanedionate, Pt 48.0% min, Thermo Scientific Chemicals
177.00 € - 750.00 €
Chemical Identifiers
CAS | 15170-57-7 |
---|---|
Molecular Formula | C10H14O4Pt |
Molecular Weight (g/mol) | 393.30 |
MDL Number | MFCD00000028 |
InChI Key | KLFRPGNCEJNEKU-FDGPNNRMSA-L |
Synonym | platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii |
PubChem CID | 10960186 |
SMILES | [Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11306048
|
Thermo Scientific Alfa Aesar
010526.03 |
1 g |
177.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396038
|
Thermo Scientific Alfa Aesar
010526.06 |
5 g |
750.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used for photoactivated hydrosilylation reactions between vinyl and silane groups; in the deposition of platinum metal and in the preparation of platinum nanotubes. Precursor for FePt superlattice magnetic nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15170-57-7 | |
393.30 | |
KLFRPGNCEJNEKU-FDGPNNRMSA-L | |
10960186 |
C10H14O4Pt | |
MFCD00000028 | |
platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii | |
[Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Specifications
250°C to 252°C | |
Red | |
C10H14O4Pt | |
platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii | |
KLFRPGNCEJNEKU-FDGPNNRMSA-L | |
platinum(2+) bis((2Z)-4-oxopent-2-en-2-olate) | |
10960186 | |
Platinum(II) 2, 4-pentanedionate, Pt 48.0% min. |
15170-57-7 | |
Powder | |
MFCD00000028 | |
Very soluble in chloroform; insoluble in water | |
[Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
393.30 | |
393.31 |
Safety and Handling
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H361
missing translation for 'einecsNumber' : 239-223-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only