missing translation for 'onlineSavingsMsg'
Learn More

Platinum gauze, 100 mesh woven from 0.0762mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

Form: approx. 0.47g/25x25mm; Wire cloth; Open area: 62.7%; Width of opening: 0.015 in.

435.17 € - 1124.60 €

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11391358
View Documents
Thermo Scientific Alfa Aesar
010282.FF
25 x 25 mm
435.17 €
Each
Please to purchase this item. Need a web account? Register with us today!
11325769
View Documents
Thermo Scientific Alfa Aesar
010282.FI
50 x 50 mm
1124.60 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

White
Silvery white guage
99.9%
MFCD00011179
Insoluble in water. Dissolves in aqua regia
[Pt]
195.08
CHEBI:33400
Odorless
Platinum
7440-06-4
21.45g/cm3 at 20°C
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
100 Mesh
Wire Cloth
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-116-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only