missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Platinum-Cured Silicone Tubing, 500 ft. spool
For pharmaceutical and biotechnology applications platinum-cured silicone tubing is an exceptional choice for fluid transfer
Brand: Masterflex™ 98906-02
This item is not returnable.
View return policy
Description
- Reduce unpacking time, save money on shipping and ensure that you always have enough tubing on hand with bulk tubing.
- Silicone provides excellent biocompatibility and chemical inertness to maintain the purity of sensitive fluids.
- Slightly greater clarity than peroxide-cured tubing.
- Smooth surfaces; lower protein binding levels.
- Fewer potential leachables.
- Ideal for pharmaceutical and biotechnology use.
Specifications
Non-Phthalate;NSF-61;NSF-51;USP;ADCF;3A;European Pharmacopoeia;FDA;Non-DEHP | |
Platinum-cured Silicone | |
500 ft. | |
Transparent |
1.6 mm | |
3.2 mm | |
0.8 mm | |
15 psi |