missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 5% on carbon paste, B501018-5, Thermo Scientific™

Product Code. 15447647
Click to view available options
Quantity:
5 g
25 g
100 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Gray
Physical Form Solid
Quantity 100 g
Odor Odorless
Chemical Name or Material Platinum, 5% on carbon paste, B501018-5
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'