missing translation for 'onlineSavingsMsg'
Learn More

Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane, Thermo Scientific Chemicals

59.50 € - 1382.88 €

Chemical Identifiers

CAS 68478-92-2
Molecular Formula C8 H18 O Pt Si2
Molecular Weight (g/mol) 474.68
MDL Number MFCD00151662
InChI Key RCNRJBWHLARWRP-UHFFFAOYSA-N
Synonym platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, platinum salt, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution, ksc628a5r, platinum 1,3-divinyl-1,1,3,3-tetramethyldisiloxane, dimethyl vinyl silyl oxy-dimethyl-vinyl-silane; platinum, ethenyl ethenyldimethylsilyl oxy dimethylsilane platinum, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, platinum,1,3-diethenyl-1,1,3,3-tetramethyldisiloxane complexes
PubChem CID 10959889
SMILES [Pt].C[Si](C)(O[Si](C)(C)C=C)C=C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314258
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041508.03
1 g
59.50 €
1g
Please to purchase this item. Need a web account? Register with us today!
11324258
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041508.06
5 g
290.45 €
5g
Please to purchase this item. Need a web account? Register with us today!
11304258
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041508.14
25 g
1382.88 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst.Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane is used as a catalyst in hydrosilylation reaction for the preparation of trans-beta-silanes and in reductive amination reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst
Specifications

Chemical Identifiers

68478-92-2
474.68
RCNRJBWHLARWRP-UHFFFAOYSA-N
10959889
C8 H18 O Pt Si2
MFCD00151662
platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, platinum salt, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution, ksc628a5r, platinum 1,3-divinyl-1,1,3,3-tetramethyldisiloxane, dimethyl vinyl silyl oxy-dimethyl-vinyl-silane; platinum, ethenyl ethenyldimethylsilyl oxy dimethylsilane platinum, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, platinum,1,3-diethenyl-1,1,3,3-tetramethyldisiloxane complexes
[Pt].C[Si](C)(O[Si](C)(C)C=C)C=C

Specifications

68478-92-2
0.984 g/mL
C8 H18 O Pt Si2
Moisture sensitive
Miscible with toluene,xylene,isopropyl alcohol,double-vinyl silicone oil and vinyl terminated poly(dimethylsiloxane). Immiscible with water.
[Pt].C[Si](C)(O[Si](C)(C)C=C)C=C
474.68
474.68
Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, Soln. in vinyl terminated polydimethylsiloxane
Orange
>110°C (230°F)
MFCD00151662
platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, platinum salt, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution, ksc628a5r, platinum 1,3-divinyl-1,1,3,3-tetramethyldisiloxane, dimethyl vinyl silyl oxy-dimethyl-vinyl-silane; platinum, ethenyl ethenyldimethylsilyl oxy dimethylsilane platinum, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, platinum,1,3-diethenyl-1,1,3,3-tetramethyldisiloxane complexes
RCNRJBWHLARWRP-UHFFFAOYSA-N
ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane;platinum
10959889
Liquid
Videos
Safety and Handling

Safety and Handling

H335

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only