Learn More
Grant Instruments™ Platform For V-32 Vortex Mixer
Creates versatility with platforms that accommodate a variety of tube sizes.
50.40 € - 54.35 €
Product Code | Brand | No. of Positions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Positions | Price | Quantity & Availability | |||||
11446367
|
Grant Instruments™
PV-32 |
32 |
54.35 €
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
15251226
|
Grant Instruments™
PV6-10 |
6 |
50.40 €
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
Description
For use with V-32 Vortex Mixer
Replacement Platform PV-32:
- Fits 3 tube sizes:
- 16 x 1.5mL
- 8 x 0.5mL
- 8 x 0.2mL
Universal 6-socket platform PV-6:
- 6 x 10mL tubes
- 15mm maximum tube dia.