missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PlasmidSelect Xtra Medium

PlasmidSelect Xtra is a thiophilic aromatic adsorption chromatography resin with a selectivity that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.

824.00 €

Specifications

For Use With (Application) Generic Purification of Supercoiled Plasmid DNA
Stability Stable in all commonly used aqueous buffers
Storage Requirements 4 to 30 deg.C, 20% Ethanol
Product Type PlasmidSelect Xtra Media
Particle Size 24 to 44 μm
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10211030
View Documents
Cytiva
28-4024-02
200 mL
824.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • PlasmidSelect Xtra allows for generic purification of supercoiled plasmid DNA
  • Allows for consistent purification results when scaling up from research scale to cGMP production
  • Convenient formats for research, process development and quantitative and qualitative analysis
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • This resin is a BioProcess medium supported for industrial applications
  • Composed of 6% highly cross-linked agarose
Specifications

Specifications

Generic Purification of Supercoiled Plasmid DNA
4 to 30 deg.C, 20% Ethanol
24 to 44 μm
<120 cm/h
3 to 11
Stable in all commonly used aqueous buffers
PlasmidSelect Xtra Media
BioProcess resin
Cross-linked Agarose
2-mercaptopyridine
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • PlasmidSelect Xtra, 200 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.