Learn More
Cowie Technology™ Cross-Shape Stirring Magnet, PTFE
A very stable general purpose stirrer. The COWIE TECHNOLOGY™ Cross-Shape Stirring Magnet has a similar action to a cylindrical stirring magnet, but gives more turbulence at low speed.
Brand:  Cowie Technology™ 001.2407
Product Code. 10620283
  • 11.65 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stirring magnet for easy and convenient mixing
  • PTFE construction
Specifications
Specifications
Stirring Magnet
3.375 in.
60 mm
PTFE