missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKM2 Mouse anti-Human, Mouse, Rat, Clone: 1C11C7, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | PKM2 |
---|---|
Clone | 1C11C7 |
Concentration | 0.53 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16847325
|
Proteintech
60268-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16837325
|
Proteintech
60268-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKM2, Pyruvate kinase isozymes M1/M2, has 2 isoforms. The immunogen of this antibody is M2 isoform, also called PKM2.
PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.Specifications
PKM2 | |
0.53 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CTHBP, OIP 3, OIP3, Opa interacting protein 3, p58, PK2, PK3, PKM, PKM1, PKM2, PKM2 pyruvate kinase, muscle, Pyruvate kinase 2/3, Pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, TCB, THBP1, Tumor M2 PK | |
PKM | |
IgG1 | |
Protein G | |
Antibody |
1C11C7 | |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Rat, Mouse | |
P11980, P14618, P52480 | |
18746, 25630, 5315 | |
Peptide | |
Primary | |
-20°C | |
PKM |