missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIWIL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15659-1-AP-20UL
This item is not returnable.
View return policy
Description
Hiwi is a member of the Piwi/Argonaute protein family. Hiwi is specifically expressed in germline cells such as spermatocytes and round spermatids during spermatogenesis. Hiwi is also expressed in CD34+ hematopoietic progenitors, but not in CD34-hematopoietic cells, suggesting a role for this gene in the maintece of hematopoietic stem cells.Specifications
PIWIL1 | |
Polyclonal | |
Unconjugated | |
PIWIL1 | |
HIWI, MIWI, PIWI, piwi like 1 (Drosophila), Piwi like protein 1, PIWIL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57749, 9271 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96J94, Q9JMB7 | |
Piwil1 | |
PIWIL1 Fusion Protein Ag8056 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |