Learn More
Pivaldehyde, 96%, Thermo Scientific Chemicals
68.34 € - 640.19 €
Chemical Identifiers
CAS | 630-19-3 |
---|---|
Molecular Formula | C5H10O |
Molecular Weight (g/mol) | 86.13 |
MDL Number | MFCD00006962 |
InChI Key | FJJYHTVHBVXEEQ-UHFFFAOYSA-N |
Synonym | trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde |
PubChem CID | 12417 |
IUPAC Name | 2,2-dimethylpropanal |
SMILES | CC(C)(C)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10354122
|
Thermo Scientific
368360050 |
5 g | Glass bottle |
68.34 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10682233
|
Thermo Scientific Acros
368360250 |
25 g | Glass bottle |
257.28 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10703803
|
Thermo Scientific Acros
368361000 |
100 g | Glass bottle |
640.19 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
630-19-3 | |
86.13 | |
FJJYHTVHBVXEEQ-UHFFFAOYSA-N | |
12417 | |
CC(C)(C)C=O |
C5H10O | |
MFCD00006962 | |
trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde | |
2,2-dimethylpropanal |
Specifications
630-19-3 | |
Colorless | |
75.0°C | |
Authentic | |
C5H10O | |
1.3775 to 1.3795 | |
01, IV, 3295 | |
15, 7624 | |
trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde | |
FJJYHTVHBVXEEQ-UHFFFAOYSA-N | |
2,2-dimethylpropanal | |
12417 | |
96% | |
Pivaldehyde, 98% |
6.0°C | |
0.7750g/mL | |
−15°C | |
95% min. (GC) | |
(CH3)3CCHO | |
MFCD00006962 | |
13,249 | |
0.775 | |
Solubility in water: negligibly soluble. | |
CC(C)(C)C=O | |
86.13 | |
86.13 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 211-134-6
- Pivaldehyde
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.