missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10873-1-AP-20UL
This item is not returnable.
View return policy
Description
PITX1 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family are involved in organ development and left-right asymmetry. This protein acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin.Specifications
PITX1 | |
Polyclonal | |
Unconjugated | |
PITX1 | |
BFT, Homeobox protein PITX1, paired like homeodomain 1, Pituitary homeobox 1, PITX1, POTX, PTX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18740, 5307 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P70314, P78337 | |
Pitx1 | |
PITX1 Fusion Protein Ag1303 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |