Learn More
Gilson™ PIPETMAN™ Classic 4-Pipette Kit
The PIPETMAN™ Classic 4-Pipette Kit provides 4 pipettes covering volume transfers from 0.2 to 1000μL, which will tackle most laboratory needs. The PIPETMAN Classic pipettes, the world standing in pipetting, are continuously adjustable air-displacement pipettes suited for aqueous solutions of moderate viscosity and density.
Brand: Gilson™ F167380
This item is not returnable.
View return policy
Description
- Provides exceptional accuracy and precision
- Easily adjusted volume can be trusted to exceed ISO recommendations when used with PIPETMAN™ Diamond tips
- Maintains consistent high level of performance for years to come with annual servicing
- PVDF body is resistant to aggressive chemicals and its piston assembly inhibits degradation of internal components
- Each kit includes: P2, P20, P200 and P1000 Classic pipettes, three TIPACK™ racks of Diamond tips, four SINGLE™ pipette holders, four PIPETMAN Comfort Handles, the Gilson™ guide to pipetting, and two-minute inspection poster
Specifications
Manual air displacement | |
Pipetman™ Classic™ | |
0.2 to 1000 μL | |
1 | |
12 Months |
Aspiration, Dispensing | |
Pipette Kit | |
Aspiration, dispensing | |
Chemical resistance |