missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIPES, disodium salt, 99%, Thermo Scientific Chemicals
56.40 € - 312.00 €
Chemical Identifiers
CAS | 76836-02-7 |
---|---|
Molecular Formula | C8H16N2Na2O6S2 |
Molecular Weight (g/mol) | 346.324 |
InChI Key | GMHSTJRPSVFLMT-UHFFFAOYSA-L |
Synonym | 1, 4-Piperazinediethanesulfonic acid disodium salt |
PubChem CID | 173553 |
ChEBI | CHEBI:63055 |
IUPAC Name | disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate |
SMILES | C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11976521
|
Thermo Scientific Acros
215090250 |
25 g | Glass bottle |
56.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11986521
|
Thermo Scientific Acros
215091000 |
100 g | Glass bottle |
154.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11996521
|
Thermo Scientific Acros
215092500 |
250 g | Glass bottle |
312.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
76836-02-7 | |
346.324 | |
1, 4-Piperazinediethanesulfonic acid disodium salt | |
CHEBI:63055 | |
C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+] |
C8H16N2Na2O6S2 | |
GMHSTJRPSVFLMT-UHFFFAOYSA-L | |
173553 | |
disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate |
Specifications
0.050 max. (0.1M in water) at 260nm, 0.050 max. (0.1M in water) at 280nm, 0.050 max. (260nm, 0.1M in water) | |
White | |
Crystalline Powder or Powder | |
Authentic | |
1, 4-Piperazinediethanesulfonic acid disodium salt | |
C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+] | |
346.324 | |
CHEBI:63055 | |
99% |
76836-02-7 | |
9.2 to 10.0 max. (25°C, 1% min. aq. soln.) | |
99% | |
C8H16N2Na2O6S2 | |
GMHSTJRPSVFLMT-UHFFFAOYSA-L | |
disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate | |
173553 | |
346.33 | |
PIPES, disodium salt |
RUO – Research Use Only