missing translation for 'onlineSavingsMsg'
Learn More

Piperidine, Pure, Fisher Chemical™

Product Code. 10528820
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 110-89-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number 5979
InChI Key NQRYJNQNLNOLGT-UHFFFAOYSA-N
Synonym hexahydropyridine, azacyclohexane, cyclopentimine, cypentil, hexazane, pentamethyleneimine, piperidin, pentamethylenimine, perhydropyridine, pentamethyleneamine
PubChem CID 8082
ChEBI CHEBI:18049
IUPAC Name piperidine
SMILES C1CCNCC1

Specifications

Color Colorless
Boiling Point 106°C
Melting Point -11°C
Formula Weight 85.15g/mol
Packaging Plastic Coated Glass bottle
Viscosity 1.46 mPaS at 20°C
Vapor Pressure 24.75mmHg at 20°C
Vapor Density 3
pH 12.6
Quantity 250 mL
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Piperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P240-Ground/bond container and receiving equipment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.