missing translation for 'onlineSavingsMsg'
Learn More

Piperazine hexahydrate, 98%, Thermo Scientific Chemicals

Product Code. 13418358
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-63-2
Molecular Formula C4H10N2·6H2O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00149389
InChI Key AVRVZRUEXIEGMP-UHFFFAOYSA-N
Synonym piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn
PubChem CID 120181
IUPAC Name piperazine;hexahydrate
SMILES C1CNCCN1.O.O.O.O.O.O

Specifications

Melting Point 41.0°C to 45.0°C
Color White
Boiling Point 145.0°C to 156.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic drum
Quantity 5 kg
Beilstein 23,4
Merck Index 14,7464
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol,practically insoluble in ether
Formula Weight 194.23
Percent Purity 98%
Physical Form Adhering Crystals
Chemical Name or Material Piperazine hexahydrate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Piperazine hexahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'