Learn More
Thermo Scientific™ Pierce™ Power Blotter
Get rapid and efficient protein transfer at your fingertips with the Thermo Scientific™ Pierce™ Power Blotter.
754.00 € - 2195.00 €
Products 3
Product Code Brand Description Includes For Use With (Application) Price Quantity & Availability  
Product Code Brand Description Includes For Use With (Application) Price Quantity & Availability  
15635315
View Documents
Thermo Scientific™
22834
Pierce Power Blotter Pierce Power Station, Pierce Power Blot Cassette, Western Blot Roller, Power Cord with C/13 Connector, Quick Start Guide Semi-dry transfer of proteins from polyacrylamide gels to membranes N/A N/A N/A
15319134
View Documents
Thermo Scientific™
22834SPCL
Pierce™ Power Blotter Welcome Pack, SDS-PAGE Gels Compatibility, Horizontal Running Dimension, Semi-Dry Mode of Transfer, 100 to 240V, 50/60Hz, 168W Electrical Requirements, Use With iBlot™, For Protein Assays and Analysis Pierce power station, pierce power blot cassette, western blot roller, power cord with C/13 connector, quick start guide, pierce 1-Step transfer buffer, 1L western blotting filter paper, 7 x 8.4cm, 100 sheets Protein assays and analysis, protein gel electrophoresis N/A N/A N/A
15399114
View Documents
Thermo Scientific™
22835
Pierce Power Blot Cassette Pierce Power Blot Cassette Semi-dry transfer when used with Pierce Power Station N/A N/A N/A
Description
Show More
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Pierce™ Power Blotter