missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic Anhydride, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
73.20 € - 133.00 €
Chemical Identifiers
CAS | 85-44-9 |
---|---|
Molecular Formula | C8H4O3 |
Molecular Weight (g/mol) | 148.12 |
MDL Number | MFCD00005918 |
InChI Key | LGRFSURHDFAFJT-UHFFFAOYSA-N |
Synonym | phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen |
PubChem CID | 6811 |
ChEBI | CHEBI:36605 |
SMILES | O=C1OC(=O)C2=CC=CC=C12 |
Chemical Identifiers
85-44-9 | |
148.12 | |
LGRFSURHDFAFJT-UHFFFAOYSA-N | |
6811 | |
O=C1OC(=O)C2=CC=CC=C12 |
C8H4O3 | |
MFCD00005918 | |
phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen | |
CHEBI:36605 |
Specifications
85-44-9 | |
C8H4O3 | |
1L = 1.53kg | |
LGRFSURHDFAFJT-UHFFFAOYSA-N | |
1,3-dihydro-2-benzofuran-1,3-dione | |
6811 | |
148.12 | |
Phthalic Anhydride |
152.00°C | |
MFCD00005918 | |
phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen | |
O=C1OC(=O)C2=CC=CC=C12 | |
148.12 | |
CHEBI:36605 | |
BAKER ANALYZED™ ACS Reagent |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.