missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) chloride, 99.99% (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 10026-13-8
Molecular Formula Cl5P
Molecular Weight (g/mol) 208.224
MDL Number MFCD00011439
InChI Key UHZYTMXLRWXGPK-UHFFFAOYSA-N
Synonym phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu
PubChem CID 24819
ChEBI CHEBI:30335
IUPAC Name pentachloro-$l^{5}-phosphane
SMILES P(Cl)(Cl)(Cl)(Cl)Cl
View More Specs

Description

Description

Phosphorus(V) chloride is used in organic synthesis, chemical reagents and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10026-13-8
208.224
UHZYTMXLRWXGPK-UHFFFAOYSA-N
24819
pentachloro-$l^{5}-phosphane
Cl5P
MFCD00011439
phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu
CHEBI:30335
P(Cl)(Cl)(Cl)(Cl)Cl

Specifications

162° (sublimation)
Yellow
99.998% (metals basis)
MFCD00011439
Insoluble in water.
P(Cl)(Cl)(Cl)(Cl)Cl
208.224
CHEBI:30335
Pungent
Yellowish crystals
2.114 g/cm3 at 20°C
10026-13-8
Solid
Cl5P
phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu
UHZYTMXLRWXGPK-UHFFFAOYSA-N
pentachloro-$l^{5}-phosphane
24819
208.24
Moisture sensitive
0.02 hPa at 20°C
Phosphorus(V) chloride
Videos
Safety and Handling

Safety and Handling

Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H330-H373

missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II

missing translation for 'einecsNumber' : 233-060-3

missing translation for 'rtecsNumber' : TB6125000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only