missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Pentoxide, Extra Pure, SLR, Fisher Chemical™

14.60 € - 45.85 €

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
IUPAC Name tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11498742
View Documents
Fisher Chemical
P/2960/48
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10468220
View Documents
Fisher Chemical
P/2960/50
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10478220
View Documents
Fisher Chemical
P/2960/53
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

1314-56-3
141.94
DLYUQMMRRRQYAE-UHFFFAOYSA-N
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
O5P2
MFCD00011440
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone

Specifications

340°C
White
Solid
MFCD00011440
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
141.94
Plastic powder jar
1314-56-3
1
O5P2
DLYUQMMRRRQYAE-UHFFFAOYSA-N
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
141.94g/mol
1.3mbar 388
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phosphorus pentoxide

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.