Learn More
Phosphorus pentachloride, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 10026-13-8 |
---|---|
Molecular Formula | Cl5P |
MDL Number | MFCD00011439 |
InChI Key | UHZYTMXLRWXGPK-UHFFFAOYSA-N |
Synonym | phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tyduShow More |
PubChem CID | 24819 |
ChEBI | CHEBI:30335 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
10026-13-8 | |
MFCD00011439 | |
phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tyduShow More | |
CHEBI:30335 |
Cl5P | |
UHZYTMXLRWXGPK-UHFFFAOYSA-N | |
24819 |
Specifications
10026-13-8 | |
Crystalline Powder and Scales | |
Cl5P | |
MFCD00011439 | |
phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu | |
P(Cl)(Cl)(Cl)(Cl)Cl | |
208.224 | |
CHEBI:30335 | |
≥97.5% | |
Phosphorus pentachloride, 98% |
Yellow | |
98% | |
PCl5 | |
01,866; 04,388; 05,534; 07,290 | |
UHZYTMXLRWXGPK-UHFFFAOYSA-N | |
pentachloro-$l^{5}-phosphane | |
24819 | |
208.24 | |
1.6g/mL |
Safety and Handling
GHS H Statement:
Fatal if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-060-3
missing translation for 'rtecsNumber' : TB6125000
missing translation for 'tsca' : TSCA
- Phosphorus pentachloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.