missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, Multi-Compendial, NF, J.T. Baker™
281.00 € - 499.00 €
Chemical Identifiers
CAS | 7664-38-2 |
---|---|
Molecular Formula | H3O4P |
Molecular Weight (g/mol) | 97.994 |
InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
Synonym | orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico |
PubChem CID | 1004 |
ChEBI | CHEBI:26078 |
IUPAC Name | phosphoric acid |
SMILES | OP(=O)(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15507804
|
Avantor J.T.Baker
0268-00 |
500 mL | PVC Coated Glass Bottle |
281.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15517804
|
Avantor J.T.Baker
0268-03 |
2.5 L | Glass Bottle |
302.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
14801332
|
Avantor J.T.Baker
0268-05 |
2.5 L | Glass Bottle, Single ship |
499.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15527804
|
Avantor J.T.Baker
0268-06 |
4 L | Poly Bottle |
399.00 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7664-38-2 | |
97.994 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
CHEBI:26078 | |
OP(=O)(O)O |
H3O4P | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
1004 | |
phosphoric acid |
Specifications
7664-38-2 | |
H3O4P | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
1004 | |
98 | |
Phosphoric Acid |
1L = 1.69kg | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
OP(=O)(O)O | |
97.994 | |
CHEBI:26078 | |
Multi-Compendial/NF |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title