missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless |
Melting Point | 21.0°C |
Boiling Point | 158.0°C |
Density | 1.6800g/mL |
Heavy Metals (as Pb) | 10ppm max. |
Packaging | Plastic drum |
Specific Gravity | 1.68 |
Quantity | 10 L |
Linear Formula | H3PO4 |
Physical Form | Liquid |
Show More |
Safety and Handling
Product Identifier
- Phosphoric acid, 85+% solution in water
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Phosphoric acid
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'