Learn More
Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals
34.90 € - 296.00 €
Chemical Identifiers
CAS | 940-71-6 |
---|---|
Molecular Formula | Cl6N3P3 |
Molecular Weight (g/mol) | 347.642 |
MDL Number | MFCD00006474 |
InChI Key | UBIJTWDKTYCPMQ-UHFFFAOYSA-N |
Synonym | phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer |
PubChem CID | 220225 |
IUPAC Name | 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene |
SMILES | N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499296
|
Thermo Scientific Alfa Aesar
A18986.06 |
5 g |
34.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409306
|
Thermo Scientific Alfa Aesar
A18986.14 |
25 g |
101.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419306
|
Thermo Scientific Alfa Aesar
A18986.22 |
100 g |
296.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
940-71-6 | |
347.642 | |
UBIJTWDKTYCPMQ-UHFFFAOYSA-N | |
220225 | |
N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl |
Cl6N3P3 | |
MFCD00006474 | |
phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer | |
2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene |
Spécification
127°C (13mmHg) | |
940-71-6 | |
MFCD00006474 | |
phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer | |
N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl | |
347.642 | |
347.66 | |
Phosphonitrilic chloride trimer |
111° to 115°C | |
Cl6N3P3 | |
UN3260 | |
UBIJTWDKTYCPMQ-UHFFFAOYSA-N | |
2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene | |
220225 | |
98% |
Sécurité et traitement
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.