Learn More
Phospho-ERK1/2 (Thr202/Tyr204) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28733-1-AP-20UL
This item is not returnable.
View return policy
Description
The Phospho-ERK1/2 (Thr202/Tyr204) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-ERK1/2 (Thr202/Tyr204). This antibody recognizes human, mouse, rat antigen. The Phospho-ERK1/2 (Thr202/Tyr204) antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
Phospho-ERK1/2 (Thr202/Tyr204) | |
Polyclonal | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
20 μL | |
5595 | |
IgG |