missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Specifications
Specifications
Concentration | 1g/L |
Packaging | Bottle |
Quantity | 250 mL |
Type | Single-component Standard Solution |
Molecular Formula | H2NaO4P |
IUPAC Name | sodium;dihydrogen phosphate |
Solvent or Matrix | H2O |
Physical Form | Liquid |
Chemical Name or Material | Phosphate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'