missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylsilane, 98%, Thermo Scientific Chemicals

Product Code. 11307885
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical and organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-32-1
Molecular Formula C9H14Si
Molecular Weight (g/mol) 150.30
MDL Number MFCD00008271
InChI Key KXFSUVJPEQYUGN-UHFFFAOYSA-N
Synonym phenyltrimethylsilane, trimethyl phenyl silane, silane, trimethylphenyl, trimethylsilyl benzene, unii-px1sg1q5yp, benzene, trimethylsilyl, silane, trimethyl phenyl, px1sg1q5yp, phenyl trimethylsilane, trimethyl-phenylsilane
PubChem CID 69849
IUPAC Name trimethyl(phenyl)silane
SMILES C[Si](C)(C)C1=CC=CC=C1

Specifications

Boiling Point 169°C to 170°C
Quantity 25 g
UN Number UN1993
Beilstein 1855012
Flash Point 40°C (104°F)
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 150.3
Percent Purity 98%
Refractive Index 1.49
Density 0.871
Chemical Name or Material Phenyltrimethylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenyltrimethylsilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Phenyltrimethylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.