missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Phenylsuccinic acid, 98%, Thermo Scientific Chemicals
27.70 € - 342.00 €
Chemical Identifiers
CAS | 635-51-8 |
---|---|
Molecular Formula | C10H10O4 |
Molecular Weight (g/mol) | 194.186 |
MDL Number | MFCD00004256 |
InChI Key | LVFFZQQWIZURIO-UHFFFAOYSA-N |
Synonym | phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl |
PubChem CID | 95459 |
IUPAC Name | 2-phenylbutanedioic acid |
SMILES | C1=CC=C(C=C1)C(CC(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443994
|
Thermo Scientific Alfa Aesar
A11971.14 |
25 g |
27.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453994
|
Thermo Scientific Alfa Aesar
A11971.22 |
100 g |
71.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463994
|
Thermo Scientific Alfa Aesar
A11971.36 |
500 g |
342.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
635-51-8 | |
194.186 | |
LVFFZQQWIZURIO-UHFFFAOYSA-N | |
95459 | |
C1=CC=C(C=C1)C(CC(=O)O)C(=O)O |
C10H10O4 | |
MFCD00004256 | |
phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl | |
2-phenylbutanedioic acid |
Specifications
635-51-8 | |
C10H10O4 | |
1875051 | |
LVFFZQQWIZURIO-UHFFFAOYSA-N | |
2-phenylbutanedioic acid | |
95459 | |
98% |
167°C to 169°C | |
MFCD00004256 | |
phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl | |
C1=CC=C(C=C1)C(CC(=O)O)C(=O)O | |
194.186 | |
194.19 | |
(±)-Phenylsuccinic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 211-238-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only