Learn More
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals
91.25 € - 531.65 €
Chemical Identifiers
CAS | 100-58-3, 5614-37-9 |
---|---|
Molecular Formula | C6H5BrMg |
Molecular Weight (g/mol) | 181.32 |
MDL Number | MFCD00000038 |
InChI Key | ANRQGKOBLBYXFM-UHFFFAOYSA-M |
Synonym | phenylmagnesium bromide, bromophenylmagnesium, magnesium, bromophenyl, phmgbr, phenylmagnesiumbromide, bromo phenyl magnesium, phenyl magnesium bromide, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, grignard reagent |
PubChem CID | 66852 |
IUPAC Name | magnesium;benzene;bromide |
SMILES | Br[Mg]C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12625017
|
Thermo Scientific Acros
445961000 |
100 mL | AcroSeal™ Glass bottle |
91.25 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12635017
|
Thermo Scientific Acros
445968000 |
800 mL | AcroSeal™ Glass bottle |
531.65 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-58-3, 5614-37-9 | |
181.32 | |
ANRQGKOBLBYXFM-UHFFFAOYSA-M | |
66852 | |
Br[Mg]C1=CC=CC=C1 |
C6H5BrMg | |
MFCD00000038 | |
phenylmagnesium bromide, bromophenylmagnesium, magnesium, bromophenyl, phmgbr, phenylmagnesiumbromide, bromo phenyl magnesium, phenyl magnesium bromide, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, grignard reagent | |
magnesium;benzene;bromide |
Specifications
100-58-3, 5614-37-9 | |
C6H5BrMg | |
MFCD00000038 | |
Solubility in water: reacts | |
Br[Mg]C1=CC=CC=C1 | |
181.32 | |
181.31 |
−1°C | |
C6H5MgBr | |
phenylmagnesium bromide, bromophenylmagnesium, magnesium, bromophenyl, phmgbr, phenylmagnesiumbromide, bromo phenyl magnesium, phenyl magnesium bromide, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, grignard reagent | |
ANRQGKOBLBYXFM-UHFFFAOYSA-M | |
magnesium;benzene;bromide | |
66852 | |
Phenylmagnesium bromide |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-867-2
- Phenylmagnesium bromide, 1.6M solution in CPME
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.