missing translation for 'onlineSavingsMsg'
Learn More

Phenyl propargyl ether, 97%, Thermo Scientific Chemicals

Product Code. 11314978
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Phenyl propargyl ether is used as building block for synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13610-02-1
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00048107
InChI Key AIQRJSXKXVZCJO-UHFFFAOYSA-N
Synonym phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl
PubChem CID 83613
IUPAC Name prop-2-ynoxybenzene
SMILES C#CCOC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.030 g/mL
Boiling Point 89°C to 90°C (14 mmHg)
Odor Pungent
Linear Formula C6H5OCH2C≡CH
Refractive Index 1.533
Quantity 100 g
Solubility Information Soluble in chloroform.
Formula Weight 132.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Phenyl propargyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'