missing translation for 'onlineSavingsMsg'
Learn More

Phenol-Indo-2,6-Dichlorophenol Sodium Salt, Pure, Fisher Chemical™

Product Code. 10305860
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Phenol-Indo-2,6-Dichlorophenol Sodium Salt, Pure, Fisher Chemical, Quantity: 5g, CAS: 620-45-1, Green, Molecular Formula: C12H6Cl2NNaO2, Molecular Weight: 290.075, Packaging: Glass Jar

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-45-1
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
PubChem CID 23697355
ChEBI CHEBI:948
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1

Specifications

Color Green
Physical Form Powder
Quantity 5 g
IUPAC Name sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Formula Weight 325.9701g/mol
Grade Pure
Packaging Glass Jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'