missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl alcohol, 99%, Thermo Scientific Chemicals

Product Code. 10562141
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-12-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002886
InChI Key WRMNZCZEMHIOCP-UHFFFAOYSA-N
Synonym phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
PubChem CID 6054
ChEBI CHEBI:49000
SMILES OCCC1=CC=CC=C1

Specifications

Melting Point -27.0°C
Color Colorless
Density 1.0200g/mL
Boiling Point 219.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2OH
Refractive Index 1.5300 to 1.5350
Quantity 2.5 L
Beilstein 06, 478
Merck Index 15, 7338
Specific Gravity 1.02
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: miscible with alcohol and ether
IUPAC Name 2-phenylethan-1-ol
Viscosity 7.58 mPa.s (25°C)
Formula Weight 122.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenethyl alcohol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'