Learn More
Phenanthridine, 98%, Thermo Scientific Chemicals
33.10 € - 300.00 €
Chemical Identifiers
CAS | 229-87-8 |
---|---|
Molecular Formula | C13H9N |
Molecular Weight (g/mol) | 179.22 |
MDL Number | MFCD00004989 |
InChI Key | RDOWQLZANAYVLL-UHFFFAOYSA-N |
Synonym | 6-phenanthridine, benzo c quinoline, 3,4-benzoquinoline, 9-azaphenanthrene, 3,4-benzoisoquinoline, 5-azaphenanthrene, unii-62qgs7cps6, ccris 1234, 62qgs7cps6 |
PubChem CID | 9189 |
ChEBI | CHEBI:36421 |
IUPAC Name | phenanthridine |
SMILES | C1=CC=C2C(=C1)C=NC3=CC=CC=C23 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10421792
|
Thermo Scientific Acros
169250010 |
1 g | Glass bottle |
33.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10194251
|
Thermo Scientific Acros
169250050 |
5 g | Glass bottle |
86.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10463785
|
Thermo Scientific Acros
169250250 |
25 g | Glass bottle |
300.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
229-87-8 | |
179.22 | |
RDOWQLZANAYVLL-UHFFFAOYSA-N | |
9189 | |
phenanthridine |
C13H9N | |
MFCD00004989 | |
6-phenanthridine, benzo c quinoline, 3,4-benzoquinoline, 9-azaphenanthrene, 3,4-benzoisoquinoline, 5-azaphenanthrene, unii-62qgs7cps6, ccris 1234, 62qgs7cps6 | |
CHEBI:36421 | |
C1=CC=C2C(=C1)C=NC3=CC=CC=C23 |
Specifications
229-87-8 | |
White to Beige | |
349.0°C (769.0 mmHg) | |
Authentic | |
C13H9N | |
20, 466 | |
Solubility in water: insoluble. Other solubilities: soluble in toluene, dmso | |
C1=CC=C2C(=C1)C=NC3=CC=CC=C23 | |
179.22 | |
CHEBI:36421 | |
98% | |
Phenanthridine |
104.0°C to 108.0°C | |
100g/mL | |
100°C | |
97.5% min. (GC) | |
MFCD00004989 | |
6-phenanthridine, benzo c quinoline, 3,4-benzoquinoline, 9-azaphenanthrene, 3,4-benzoisoquinoline, 5-azaphenanthrene, unii-62qgs7cps6, ccris 1234, 62qgs7cps6 | |
RDOWQLZANAYVLL-UHFFFAOYSA-N | |
phenanthridine | |
9189 | |
179.22 | |
Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Suspected of causing genetic defects.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-934-4
- Phenanthridine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only