missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ pH Electrode
This unique maintenance free solid electrolyte pH electrode for measurement up to 110 ° C and 10 bar, is ideal for industrial applications.
82.80 € - 331.00 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11759748
|
Fisherbrand™
1932527-010B |
102 mm | N/A | ||||||
11785628
|
Fisherbrand™
3917871-010B |
115 mm |
331.00 €
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
11755618
|
Fisherbrand™
BP635-DSP-B120 |
120 mm |
161.00 €
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
11765618
|
Fisherbrand™
HABP635-DSP-225 |
225 mm |
213.00 €
Each |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
11739738
|
Fisherbrand™
P721P3000309AB |
152 mm |
228.00 €
Each |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
Description
- Maintenance free solid electrolyte pH electrode for measurement up to 110 ° C and 10 bar
- Autoclavable 121 ° C
- Length 12 or 22.5 cm
- External thread PG 13,5 for in-line mounting
- Ideal for industrial applications