missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 3.00 Buffer Solution at 25°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
26.45 € - 87.45 €
Specifications
Description | Accuracy ± 0.02 pH units |
---|---|
pH | 3.00 |
Physical Form | Liquid |
Packaging | HDPE Bottle GL45 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484539
|
Reagecon
1030525 |
500 mL |
87.45 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494539
|
Reagecon
103025 |
1 L |
26.45 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Spécification
Accuracy ± 0.02 pH units | |
3.00 | |
Liquid | |
HDPE Bottle GL45 |